COM448668538 Combi-Blocks, Inc.

4-[3-(Chloromethyl)phenyl]carbonylmorpholine

  • MDL number MFCD11208347
  • Cas Number : 1094300-44-3
  • Molecular Weight : 239.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4076
Availability details for
Quantity
  • Availability
  • Add to Cart