COM448668564 Combi-Blocks, Inc.

1-ethoxy-2,3-dimethylbenzene

  • MDL number MFCD28096652
  • Cas Number : 61808-02-4
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4119
Availability details for
Quantity
  • Availability
  • Add to Cart