COM448668565 Combi-Blocks, Inc.

1,2-Dimethyl-3-propoxybenzene

  • MDL number MFCD26386335
  • Cas Number : 833460-92-7
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4120

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4120
Availability details for
Quantity
  • Availability
  • Add to Cart