COM448668568 Combi-Blocks, Inc.

3-[(Diethylamino)methyl]benzonitrile

  • MDL number MFCD09931537
  • Cas Number : 1016701-33-9
  • Molecular Weight : 188.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4127

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4127
Availability details for
Quantity
  • Availability
  • Add to Cart