COM448668577 Combi-Blocks, Inc.

2-Iodo-N,N-dimethylbenzamide

  • MDL number MFCD00463660
  • Cas Number : 54616-46-5
  • Molecular Weight : 275.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4136
Availability details for
Quantity
  • Availability
  • Add to Cart