COM448668591 Combi-Blocks, Inc.

3-Iodo-N,N-dimethylbenzamide

  • MDL number MFCD02859819
  • Cas Number : 91506-06-8
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4151

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4151
Availability details for
Quantity
  • Availability
  • Add to Cart