COM448668596 Combi-Blocks, Inc.

1-[(4-Iodophenyl)carbonyl]pyrrolidine

  • MDL number MFCD00684252
  • Cas Number : 168317-99-5
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4156

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4156
Availability details for
Quantity
  • Availability
  • Add to Cart