COM448668597 Combi-Blocks, Inc.

N-Benzyl-4-iodobenzamide

  • MDL number MFCD01014865
  • Cas Number : 179117-56-7
  • Molecular Weight : 337.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4157

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4157
Availability details for
Quantity
  • Availability
  • Add to Cart