COM448668605 Combi-Blocks, Inc.

N-Benzyl-3-iodobenzamide

  • MDL number MFCD02859811
  • Cas Number : 82082-51-7
  • Molecular Weight : 337.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4166
Availability details for
Quantity
  • Availability
  • Add to Cart