COM448668612 Combi-Blocks, Inc.

1,2-Dichloro-3-isopropoxybenzene

  • MDL number MFCD23114217
  • Cas Number : 197634-45-0
  • Molecular Weight : 205.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4174
Availability details for
Quantity
  • Availability
  • Add to Cart