COM448668614 Combi-Blocks, Inc.

1-(4-Carboxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD12207487
  • Cas Number : 40306-01-2
  • Molecular Weight : 249.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4182

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4182
Availability details for
Quantity
  • Availability
  • Add to Cart