COM448668631 Combi-Blocks, Inc.

4-Cyano-N-phenylbenzamide

  • MDL number MFCD00505998
  • Cas Number : 17922-96-2
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4206

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4206
Availability details for
Quantity
  • Availability
  • Add to Cart