COM448668634 Combi-Blocks, Inc.

6-Mprpholinopicoline

  • MDL number MFCD28369599
  • Cas Number : 1321517-90-1
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4223

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4223
Availability details for
Quantity
  • Availability
  • Add to Cart