COM448668639 Combi-Blocks, Inc.

3-Cyano-N-isopropylbenzamide

  • MDL number MFCD09940478
  • Cas Number : 623569-57-3
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4228
Availability details for
Quantity
  • Availability
  • Add to Cart