COM448668759 Combi-Blocks, Inc.

4-(Benzyloxy)-3-bromopyridine

  • MDL number MFCD11226996
  • Cas Number : 1019767-63-5
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1175

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1175
Availability details for
Quantity
  • Availability
  • Add to Cart