COM448668984 Combi-Blocks, Inc.

6-Bromo-3-iodopyridin-2-ol

  • MDL number MFCD27981326
  • Cas Number : 1820674-50-7
  • Molecular Weight : 299.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1539

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1539
Availability details for
Quantity
  • Availability
  • Add to Cart