COM448669061 Combi-Blocks, Inc.

Methyl 6-methoxypyrimidine-4-carboxylate

  • MDL number MFCD27936853
  • Cas Number : 1244019-84-8
  • Molecular Weight : 168.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1784

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1784
Availability details for
Quantity
  • Availability
  • Add to Cart