COM448669063 Combi-Blocks, Inc.

tert-Butyl 4-oxo-3H-pyrrolo[2,3-d]pyrimidine-7-carboxylate

  • MDL number MFCD27981328
  • Cas Number : 1820739-67-0
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1787

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1787
Availability details for
Quantity
  • Availability
  • Add to Cart