COM448669068 Combi-Blocks, Inc.

6-Chloro-5-fluoro-2-methoxypyridine-3-carbonitrile

  • MDL number MFCD27981329
  • Cas Number : 1820650-64-3
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1795

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1795
Availability details for
Quantity
  • Availability
  • Add to Cart