COM448669069 Combi-Blocks, Inc.

tert-Butyl 6-bromo-1H-pyrrolo[3,2-b]pyridine-1-carboxylate

  • MDL number MFCD27981330
  • Cas Number : 1820711-82-7
  • Molecular Weight : 297.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1799

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1799
Availability details for
Quantity
  • Availability
  • Add to Cart