COM448669070 Combi-Blocks, Inc.

2-[(Thiazolidin-2-ylidene)aminomethylidene]malonic acid

  • MDL number MFCD27981331
  • Cas Number : 1820748-73-9
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1800

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1800
Availability details for
Quantity
  • Availability
  • Add to Cart