COM448669071 Combi-Blocks, Inc.

5-Bromo-4-ethyl-3,4-dihydropyrimidine

  • MDL number MFCD20681885
  • Cas Number : 224429-98-5
  • Molecular Weight : 189.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1801
Availability details for
Quantity
  • Availability
  • Add to Cart