COM448669080 Combi-Blocks, Inc.

3-Bromopyridine-2-carboxylic acid hydrochloride

  • MDL number MFCD27981334
  • Cas Number : 1403967-63-4
  • Molecular Weight : 238.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1815

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1815
Availability details for
Quantity
  • Availability
  • Add to Cart