COM448669081 Combi-Blocks, Inc.

N-(3-Formylpyridin-2-yl)acetamide

  • MDL number MFCD17170608
  • Cas Number : 54856-85-8
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1816

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1816
Availability details for
Quantity
  • Availability
  • Add to Cart