COM448669082 Combi-Blocks, Inc.

N-(5-Bromo-2-hydroxypyridin-3-yl)formamide

  • MDL number MFCD27981335
  • Cas Number : 1820683-69-9
  • Molecular Weight : 217.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1820

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1820
Availability details for
Quantity
  • Availability
  • Add to Cart