COM448669083 Combi-Blocks, Inc.

3-Bromo-5-chloro-2-methylpyridine

  • MDL number MFCD12911787
  • Cas Number : 131036-39-0
  • Molecular Weight : 206.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1822

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1822
Availability details for
Quantity
  • Availability
  • Add to Cart