COM448669086 Combi-Blocks, Inc.

1,3-Bis(5-bromo-3-methylpyridin-2-yl)urea

  • MDL number MFCD18324111
  • Cas Number : 1263208-76-9
  • Molecular Weight : 400.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1825

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1825
Availability details for
Quantity
  • Availability
  • Add to Cart