COM448669088 Combi-Blocks, Inc.

6-Chloro-3-iodo-2-methoxypyridine

  • MDL number MFCD18415726
  • Cas Number : 1261769-34-9
  • Molecular Weight : 269.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1827

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1827
Availability details for
Quantity
  • Availability
  • Add to Cart