COM448669092 Combi-Blocks, Inc.

1-Phenylpyrazole-4-sulfonic acid

  • MDL number MFCD27981337
  • Cas Number : 18336-36-2
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1836

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1836
Availability details for
Quantity
  • Availability
  • Add to Cart