COM448669094 Combi-Blocks, Inc.

4-Isopropyl-2-methoxypyridine

  • MDL number MFCD28014044
  • Cas Number : 1370633-62-7
  • Molecular Weight : 151.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1843
Availability details for
Quantity
  • Availability
  • Add to Cart