COM448669098 Combi-Blocks, Inc.

5-Amino-3-chloropicoline

  • MDL number MFCD11040270
  • Cas Number : 896161-13-0
  • Molecular Weight : 142.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1849

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1849
Availability details for
Quantity
  • Availability
  • Add to Cart