COM448669101 Combi-Blocks, Inc.

5-Chloroquinazolin-4-amine

  • MDL number MFCD02323641
  • Cas Number : 19808-34-5
  • Molecular Weight : 179.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1854
Availability details for
Quantity
  • Availability
  • Add to Cart