COM448669114 Combi-Blocks, Inc.

Methyl 3-pivalamidoisonicotinate

  • MDL number MFCD28122696
  • Cas Number : 1820642-01-0
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1875

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1875
Availability details for
Quantity
  • Availability
  • Add to Cart