COM448669119 Combi-Blocks, Inc.

Dimethyl[(E)-2-(3-nitropyridin-2-yl)ethenyl]amine

  • MDL number MFCD11054129
  • Cas Number : 343569-94-8
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1884

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1884
Availability details for
Quantity
  • Availability
  • Add to Cart