COM448669128 Combi-Blocks, Inc.

7-Bromo-2-(di-t-butoxycarbonylamino)quinazoline

  • MDL number MFCD28101652
  • Cas Number : 1820683-11-1
  • Molecular Weight : 424.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1900

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1900
Availability details for
Quantity
  • Availability
  • Add to Cart