COM448669129 Combi-Blocks, Inc.

2-Bromo-5-(cyclopropylmethoxy)pyridine

  • MDL number MFCD22690535
  • Cas Number : 1177269-06-5
  • Molecular Weight : 228.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1901

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1901
Availability details for
Quantity
  • Availability
  • Add to Cart