COM448669137 Combi-Blocks, Inc.

2-[(2-Chloropyridin-3-yl)oxy]ethanol

  • MDL number MFCD21120525
  • Cas Number : 313657-79-3
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1911

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1911
Availability details for
Quantity
  • Availability
  • Add to Cart