COM448669138 Combi-Blocks, Inc.

2-[2-(Benzyloxy)ethoxy]pyridin-3-amine

  • MDL number MFCD28122698
  • Cas Number : 1820665-93-7
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1912

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1912
Availability details for
Quantity
  • Availability
  • Add to Cart