COM448669140 Combi-Blocks, Inc.

Methyl 2-phenoxynicotinate

  • MDL number MFCD00100674
  • Cas Number : 36701-88-9
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1914

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1914
Availability details for
Quantity
  • Availability
  • Add to Cart