COM448669142 Combi-Blocks, Inc.

2-Bromo-5-(pyrrolidin-1-yl)pyridine

  • MDL number MFCD11869570
  • Cas Number : 1142197-42-9
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1916

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1916
Availability details for
Quantity
  • Availability
  • Add to Cart