COM448669147 Combi-Blocks, Inc.

3-(2-Hydroxyethoxy)pyridin-2-ol

  • MDL number MFCD28127198
  • Cas Number : 1820648-06-3
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1927
Availability details for
Quantity
  • Availability
  • Add to Cart