COM448669148 Combi-Blocks, Inc.

N,3-Dimethylpyridine-2-carboxamide

  • MDL number MFCD19671808
  • Cas Number : 1037045-67-2
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1930

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1930
Availability details for
Quantity
  • Availability
  • Add to Cart