COM448669149 Combi-Blocks, Inc.

N,N,3-Trimethylpyridine-2-carboxamide

  • MDL number MFCD20366305
  • Cas Number : 1341499-43-1
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1931
Availability details for
Quantity
  • Availability
  • Add to Cart