COM448669152 Combi-Blocks, Inc.

2-Methanesulfonylpyridine-4-carboxamide

  • MDL number MFCD28127199
  • Cas Number : 1820707-92-3
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1937

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1937
Availability details for
Quantity
  • Availability
  • Add to Cart