COM448669155 Combi-Blocks, Inc.

N-(6-Bromo-[1,3]thiazolo[5,4-b]pyridin-2-yl)benzamide

  • MDL number MFCD11109829
  • Cas Number : 588730-01-2
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1942

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1942
Availability details for
Quantity
  • Availability
  • Add to Cart