COM448669160 Combi-Blocks, Inc.

2-Chloro-5-(1-ethyl-3,6-dihydro-2H-pyridin-4-yl)-3-methylpyridine

  • MDL number MFCD28127200
  • Cas Number : 1820717-84-7
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1960

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1960
Availability details for
Quantity
  • Availability
  • Add to Cart