COM448669161 Combi-Blocks, Inc.

2-(5-Bromo-3-methylpyridin-2-yl)propanedinitrile

  • MDL number MFCD28127201
  • Cas Number : 1820712-23-9
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1961

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1961
Availability details for
Quantity
  • Availability
  • Add to Cart