COM448669165 Combi-Blocks, Inc.

3-Bromo-5-chloro-2-methoxy-4-methylpyridine

  • MDL number MFCD17014980
  • Cas Number : 851607-30-2
  • Molecular Weight : 236.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1969

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1969
Availability details for
Quantity
  • Availability
  • Add to Cart