COM448669166 Combi-Blocks, Inc.

3-Methoxypyridin-2-ol hydrochloride

  • MDL number MFCD28127203
  • Cas Number : 1820686-01-8
  • Molecular Weight : 161.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1972

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1972
Availability details for
Quantity
  • Availability
  • Add to Cart