COM448669167 Combi-Blocks, Inc.

tert-Butyl 6-formyl-3,4-dihydro-2H-quinoline-1-carboxylate

  • MDL number MFCD11858409
  • Cas Number : 1187933-34-1
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 96
Storage temp. Refrigerated

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1973
Availability details for
Quantity
  • Availability
  • Add to Cart